Learn More
Sigma Aldrich Fine Chemicals Biosciences Round crystal window NaCl1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1235951EA
Encompass
Very soft water soluble crystal low cost and good transmission range fogs.
Catalog No. 50-289-9990