Learn More
Sigma Aldrich Fine Chemicals Biosciences Round crystal window NaCl1EA
Catalog No. 502900390
Encompass
Very soft water soluble crystal low cost and good transmission range fogs.
Catalog No. 50-290-0390 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2677241EA
Very soft water soluble crystal low cost and good transmission range fogs.