Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Cylindrical HDPE Reagent Bottle
SDP

Catalog No. 03084204 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Quantity:
230/Cs.
24/Cs.
77/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
03-084-204 1000 mL 77/Cs.
03-084-202 500 mL 24/Cs.
03-084-201 250 mL 230/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-084-204 Supplier DWK Life Sciences Supplier No. W221183
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Cylindrical HDPE Reagent Bottle is a round, narrow-mouth plastic bottle made from natural-color HDPE with good chemical resistance. Comes with a pre-attached white polypropylene screw cap with a foamed polyethylene liner.

  • Made from High Density Polyethylene (HDPE) in natural color
  • Good chemical resistance for a wide range of laboratory reagents
  • Narrow mouth design ideal for liquids
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner
  • Cylindrical round bottle profile for stable bench storage
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Natural
Material HDPE
Quantity 77/Cs.
Sterility Non-sterile
Closure Color White
Closure Included Yes
Graduated No
Mouth Narrow
Closure Liner Polyethylene
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 34 oz.
Shape Round
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.