Learn More
MSC PRO-SAFE Drum Liner, LDPE, 55 gal, 10 mil, 40 Pack
Catalog No. 502886354
Encompass
PRO-SAFE drum liners are engineered for efficient and secure containment within 55 gal. drums. Constructed from durable LDPE material, these flexible liners provide reliable protection for various industrial and laboratory contents.
  • Compatible with 55 gal. drums
  • Constructed from LDPE material
  • Measures 38 in diameter and 56 in high
  • Features a 10 mil thickness
  • Flexible drum liner style
  • Non-antistatic
  • Supplied in a 40-pack
Catalog No. 50-288-6354 Supplier MSC Supplier No. 76010594
PRO-SAFE drum liners are engineered for efficient and secure containment within 55 gal. drums. Constructed from durable LDPE material, these flexible liners provide reliable protection for various industrial and laboratory contents.
  • Compatible with 55 gal. drums
  • Constructed from LDPE material
  • Measures 38 in diameter and 56 in high
  • Features a 10 mil thickness
  • Flexible drum liner style
  • Non-antistatic
  • Supplied in a 40-pack