Learn More
Grainger Young Bros. Stamp Works Letter and Number Set, Steel, 3/8 in., 36/Set
Catalog No. 502788076
Encompass_Preferred
This combination letter and number set features precision-engineered steel stamps designed to produce low-stress impressions. The rounded character design helps prevent material fracture, making these tools suitable for critical applications in industries such as aerospace, defense, energy, and oil. Each stamp is constructed from durable steel and hardened to 50 HRC to ensure consistent performance when marking surfaces with a maximum hardness of 50 Rockwell C.
  • Includes 36 pieces comprising uppercase letters A to Z, numbers 0 to 8 (with 6 serving as 9), and an ampersand (&)
  • Rounded-face character profile minimizes stress and risk of surface fracture
  • Shank size is 9/16 in. square with a length of 3-1/8 in.
  • Hardness rating of 50 HRC
  • Suitable for industrial marking applications
Catalog No. 50-278-8076 Supplier Grainger Supplier No. 18G347
This combination letter and number set features precision-engineered steel stamps designed to produce low-stress impressions. The rounded character design helps prevent material fracture, making these tools suitable for critical applications in industries such as aerospace, defense, energy, and oil. Each stamp is constructed from durable steel and hardened to 50 HRC to ensure consistent performance when marking surfaces with a maximum hardness of 50 Rockwell C.
  • Includes 36 pieces comprising uppercase letters A to Z, numbers 0 to 8 (with 6 serving as 9), and an ampersand (&)
  • Rounded-face character profile minimizes stress and risk of surface fracture
  • Shank size is 9/16 in. square with a length of 3-1/8 in.
  • Hardness rating of 50 HRC
  • Suitable for industrial marking applications