Learn More
Electron Microscopy Sciences Round, Foam-Tip Swab on a 6" long wooden shaft. Head size:⅜" (D) x 1-3/16" (L), 50/pk
SDP
Catalog No. 50949169
Encompass_Preferred
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Foam tip over regular-cotton. 6" long wooden shaft. Head size: 38"Dia x 1316"L
Catalog No. 50-949-169 Supplier Electron Microscopy Sciences Supplier No. 72312
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Foam tip over regular-cotton. 6" long wooden shaft. Head size: 38"Dia x 1316"L