Learn More
Ted Pella Inc ROUND GLASS COVERSLIP 18MM/1OZ
SDP
Catalog No. NC9475071
Encompass
Round Glass Coverslips made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in 1oz packaging. Not sterile. 18mm dia.
Catalog No. NC9475071 Supplier Ted Pella Inc Supplier No. 26022
Round Glass Coverslips made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in 1oz packaging. Not sterile. 18mm dia.