Learn More
Electron Microscopy Sciences TEM Round Grid Mats
SDP
Catalog No. 50930879
Encompass_Preferred
White silicone rubber mats, with numbered compartments. Good for organizing grids. They will not slide or jump between compartments. Also ideal for staining grids. Easy to pick up grids without damaging forcep tips.
Catalog No. 50-930-879 Supplier Electron Microscopy Sciences Supplier No. 71170
White silicone rubber mats, with numbered compartments. Good for organizing grids. They will not slide or jump between compartments. Also ideal for staining grids. Easy to pick up grids without damaging forcep tips.