Learn More
Fine Science Tools ROUND HANDLED SUTURE
SDP
Catalog No. NC0777962
Encompass
Product: Round Handled Suture Tying Forceps/ Tip Dimensions: 0.35 x 0.35mm/ Platform Length: 5.5mm/ Alloy: Stainless Steel/ Length: 10.5cm/ Unit: Each
Catalog No. NC0777962 Supplier Fine Science Tools Supplier No. 1802610
Product: Round Handled Suture Tying Forceps/ Tip Dimensions: 0.35 x 0.35mm/ Platform Length: 5.5mm/ Alloy: Stainless Steel/ Length: 10.5cm/ Unit: Each