Learn More
Med Vet International Round Hole Plate 2.7mm, 5 holes, 30mm, J0405D
Catalog No. 5000582771
Encompass_Preferred
6mm W x 2mm thick bone plates for 2.7mm (7/64") screws. The 2.7mm round-hole plate has holes at 6mm intervals, and comes in a range of lengths.
Catalog No. 50-005-82771 Supplier Med Vet International Supplier No. JOR405D
6mm W x 2mm thick bone plates for 2.7mm (7/64") screws. The 2.7mm round-hole plate has holes at 6mm intervals, and comes in a range of lengths.