Learn More
Med Vet International Round Hole Plate 3.5mm, 9 holes, 106mm, J0407K
Catalog No. 5000582305
Encompass_Preferred
A stronger and less expensive alternative to 3.5mm compression plates. Material is compatible with your choice of cortical or self-tapping screws.
Catalog No. 50-005-82305 Supplier Med Vet International Supplier No. JOR407K
A stronger and less expensive alternative to 3.5mm compression plates. Material is compatible with your choice of cortical or self-tapping screws.