Learn More
Med Vet International Round-Hole Plate, Heavy-Duty, J0408D, 3.5mm, 11 holes, 143mm, Each
Catalog No. 5000582630
Encompass_Preferred
12mm W, 4mm thick heavy-duty plate is stronger than either the 3.5mm compression plate or our 3.5mm standard round-hole plate. Use of this plate makes 3.5mm cortical or 9/64" screws appropriate for the larger canine breeds.
Catalog No. 50-005-82630 Supplier Med Vet International Supplier No. JOR408D
12mm W, 4mm thick heavy-duty plate is stronger than either the 3.5mm compression plate or our 3.5mm standard round-hole plate. Use of this plate makes 3.5mm cortical or 9/64" screws appropriate for the larger canine breeds.