Learn More
USA Scientific Inc Tough-Spots, 3/8 inch diameter, white, on rolls
Catalog No. NC9281560
Encompass
Tough-Spots, small, white labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C
Catalog No. NC9281560 Supplier USA Scientific Inc Supplier No. 91850000
Tough-Spots, small, white labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C