Learn More
Electron Microscopy Sciences Round Object Holder Microtome/Cryostat, 25 mm
SDP
Catalog No. 5030336
Encompass
Machined from solid aluminum with an embossed surface for holding frozen embedding blocks. Measures: 26 mm top diameter (without ʽO' ring), 28.55 mm diameter with ʽO' ring, 23 mm body diameter, 16 mm height .
Catalog No. 50-303-36 Supplier Electron Microscopy Sciences Supplier No. 6253701
Machined from solid aluminum with an embossed surface for holding frozen embedding blocks. Measures: 26 mm top diameter (without ʽO' ring), 28.55 mm diameter with ʽO' ring, 23 mm body diameter, 16 mm height .