Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Lange™ Rectangular Plastic Disposable Cuvettes

Catalog No. 501997881 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Path Length:
50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Path Length
50-199-7881 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-7881 Supplier Hach Lange™ Supplier No. LZM130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for spectral measurements of liquids. Hach Lange™ Rectangular Plastic Disposable Cuvettes are important accessories for spectrophotometers.

  • Rectangular
  • Disposable
  • 50x10 mm
  • 50 pcs
  • Plastic
TRUSTED_SUSTAINABILITY

Specifications

Type Cuvette
Disposable Single-use
Material Plastic
Length (Metric) 50cm
Quantity 50Pack
Path Length 50 mm
For Use With (Equipment) LICO Spectrophotometer
For Use With (Application) Spectral Measurement of Liquids
Shape Rectangle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.