Learn More
Grainger Round Rod Aluminum 36 in Overall L PK3
Catalog No. 502764529
Encompass_Preferred
The Aluminum Rod 6061 from Grainger is 3/8 inch in diameter and is made of aluminum alloy 6061. It has excellent corrosion resistance and good machinability and surface finishing. The rod has a tensile strength range of 38-45 ksi and meets ASTM B211 specifications. It comes in 12 foot lengths and has a smooth surface finish. The aluminum rod can be used for shafts, fixtures, couplings and other precision parts where strength and corrosion resistance is needed.
Catalog No. 50-276-4529 Supplier Grainger Supplier No. 16NJ04
The Aluminum Rod 6061 from Grainger is 3/8 inch in diameter and is made of aluminum alloy 6061. It has excellent corrosion resistance and good machinability and surface finishing. The rod has a tensile strength range of 38-45 ksi and meets ASTM B211 specifications. It comes in 12 foot lengths and has a smooth surface finish. The aluminum rod can be used for shafts, fixtures, couplings and other precision parts where strength and corrosion resistance is needed.