Learn More
Grainger Round Rod SS 36 in Overall L PK3
Catalog No. 502764612
Encompass_Preferred
The Stainless Steel Rod 303 is a 3/16" diameter, 36" long rod made of 303 stainless steel with a smooth surface finish. It offers excellent corrosion resistance, durability, and can withstand temperatures up to 1600°F. Meeting ASTM A276 specifications, it has a tensile strength of 85,000 psi. Commonly used for shafts, fixtures, pins, and linkages, it provides good machinability and weldability. The rod comes with a mill certificate verifying its chemistry and physical properties and is available in full 36" lengths.
Catalog No. 50-276-4612 Supplier Grainger Supplier No. 16NJ07
The Stainless Steel Rod 303 is a 3/16" diameter, 36" long rod made of 303 stainless steel with a smooth surface finish. It offers excellent corrosion resistance, durability, and can withstand temperatures up to 1600°F. Meeting ASTM A276 specifications, it has a tensile strength of 85,000 psi. Commonly used for shafts, fixtures, pins, and linkages, it provides good machinability and weldability. The rod comes with a mill certificate verifying its chemistry and physical properties and is available in full 36" lengths.