Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ROUND STICKRS 3/4 GRN 1008/PK

Catalog No. 501909538
Encompass

These removable color-coding labels are ideal for organizing documents, inventory, and sale items in various environments. They feature a removable adhesive, allowing for easy repositioning or removal, and are compatible with both laser and inkjet printers for custom printing or can be written on by hand.

  • Removable adhesive
  • Green color
  • Has a 3/4 in diameter
  • Contains 1,008 labels total
  • Paper label material
  • For use with laser or inkjet printers
  • Customizable with free templates
  • Aids in organizing home or workplace

Catalog No. 50-190-9538 Supplier MSC Supplier No. 71376321
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These removable color-coding labels are ideal for organizing documents, inventory, and sale items in various environments. They feature a removable adhesive, allowing for easy repositioning or removal, and are compatible with both laser and inkjet printers for custom printing or can be written on by hand.

  • Removable adhesive
  • Green color
  • Has a 3/4 in diameter
  • Contains 1,008 labels total
  • Paper label material
  • For use with laser or inkjet printers
  • Customizable with free templates
  • Aids in organizing home or workplace

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.