Learn More
StatLab Round Tissue Water Bath Lid
SDP
Catalog No. 21060047
Change view
Click to view available options
Type:
Flat Cover
Catalog No. Type
21-060-047 Flat Cover
1 options
Catalog No. 21-060-047 Supplier StatLab Supplier No. SITWBRNDLID
Optional, easy to use drop-on water bath lid improves heating efficiency and control.
  • Easy to use drop-on water bath lid improves heating efficiency and control, while protecting water from contamination
  • Lid only, water bath sold separately
  • Intended for use with StatLab Digital Tissue Water Bath, item number SI-PWD-LG
Specifications
Type Flat Cover