Learn More
Quantum Storage Systems Round Utility Container, Crusader, Gray, 20 Gallon
Catalog No. NC3825072
Encompass_Preferred
Crusader Round Heavy-duty Utility Container, 20 Gallon, 19-1/2"""" Dia. x 22-9/10"""" H, Polyethylene, Gray, NSF, Made in USA
Catalog No. NC3825072 Supplier Quantum Storage Systems Supplier No. CTC20GY
Crusader Round Heavy-duty Utility Container, 20 Gallon, 19-1/2"""" Dia. x 22-9/10"""" H, Polyethylene, Gray, NSF, Made in USA