Learn More
Quantum Storage Systems Round Utility Container, Crusader, White, 44 Gallon
Catalog No. NC3825124
Encompass_Preferred
Crusader Round Heavy-duty Utility Container, 44 Gallon, 24"""" Dia. x 31-1/2"""" H, Polyethylene, White, NSF, Made in USA
Catalog No. NC3825124 Supplier Quantum Storage Systems Supplier No. CTC44WT
Crusader Round Heavy-duty Utility Container, 44 Gallon, 24"""" Dia. x 31-1/2"""" H, Polyethylene, White, NSF, Made in USA