Learn More
Quantum Storage Systems Round Utility Container, Crusader, Yellow, 44 Gallon
Catalog No. NC3825131
Encompass_Preferred
Crusader Round Heavy-duty Utility Container, 44 Gallon, 24"""" Dia. x 31-1/2"""" H, Polyethylene, Yellow, NSF, Made in USA
Catalog No. NC3825131 Supplier Quantum Storage Systems Supplier No. CTC44YL
Crusader Round Heavy-duty Utility Container, 44 Gallon, 24"""" Dia. x 31-1/2"""" H, Polyethylene, Yellow, NSF, Made in USA