Learn More
Quantum Storage Systems Round Utility Containers Parts and Accessories, Crusader, Round Container Lid, Fits 10 Gallon, Red
Catalog No. NC3825171
Encompass_Preferred
Crusader Round Heavy-duty Utility Container Lid, 15-3/5"""" Dia., for 10 Gallon trash can, Polyethylene, Red, NSF, Made in USA
Catalog No. NC3825171 Supplier Quantum Storage Systems Supplier No. CTCL10RD
Crusader Round Heavy-duty Utility Container Lid, 15-3/5"""" Dia., for 10 Gallon trash can, Polyethylene, Red, NSF, Made in USA