Learn More
Quantum Storage Systems Round Utility Containers Parts and Accessories, Crusader, Round Container Lid, Fits 44 Gallon, Red
Catalog No. NC3826753
Encompass_Preferred
Crusader Round Heavy-duty Utility Container Lid, 24"""" Dia., for 44 Gallon trash can, Polyethylene, Red, NSF, Made in USA
Catalog No. NC3826753 Supplier Quantum Storage Systems Supplier No. CTCL44RD
Add to Cart
Add to Cart
Crusader Round Heavy-duty Utility Container Lid, 24"""" Dia., for 44 Gallon trash can, Polyethylene, Red, NSF, Made in USA