Learn More
Ampco Round Wire Push Broom
SDP
Catalog No. 19174250 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
12 in. Length
Catalog No. Length (English)
19-174-250 12 in. Length
1 options
Catalog No. 19-174-250 Supplier Ampco Supplier No. PB10
Add to Cart
Add to Cart
Frequently purchased with Ampco dust pans. Handle is sold separately. Made in the USA.
  • Handle not included
  • See image for additional information regarding measurements.
  • Bristle length (a): 2 and 1/2 in.
  • Broom head length (b): 12 in.
  • Bristle total width (c): 2 and 1/2 in.
Specifications
Product Type Round Wire Broom
Length (English) 12 in. Length
Quantity 1 Ea.