Learn More
Grainger Roundover Cut Profile Router Bit 1-1/2
Catalog No. 502765221
Encompass_Preferred
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank carbide tipped router bit that creates a rounded edge profile. It has a cutting diameter of 1-1/4", a cutting length of 1/2", and an overall length of 2-1/2". This two flute spiral design router bit is designed for use on wood, plywood, and laminates. It can be used for decorative edging or easing sharp edges. The carbide tips provide a long service life. This is a high quality router bit made in the USA.
Catalog No. 50-276-5221 Supplier Grainger Supplier No. 16Y568
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank carbide tipped router bit that creates a rounded edge profile. It has a cutting diameter of 1-1/4", a cutting length of 1/2", and an overall length of 2-1/2". This two flute spiral design router bit is designed for use on wood, plywood, and laminates. It can be used for decorative edging or easing sharp edges. The carbide tips provide a long service life. This is a high quality router bit made in the USA.