Learn More
Grainger Roundover Cut Profile Router Bit 3/4
Catalog No. 502767166
Encompass_Preferred
The WESTWARD Roundover Cut Profile Router Bit is a 1/4" shank roundover bit with a 1/4" cutting diameter and 1/8" cutting radius. It is made of high-speed steel and has 2 flutes. This bit is designed for use in handheld or table-mounted routers and creates a rounded profile edge on wood, plastic, and soft metals. The roundover bit can be used to ease sharp edges on furniture, cabinets, crafts, and other woodworking projects. This is a high quality American-made router bit.
Catalog No. 50-276-7166 Supplier Grainger Supplier No. 16Y565
The WESTWARD Roundover Cut Profile Router Bit is a 1/4" shank roundover bit with a 1/4" cutting diameter and 1/8" cutting radius. It is made of high-speed steel and has 2 flutes. This bit is designed for use in handheld or table-mounted routers and creates a rounded profile edge on wood, plastic, and soft metals. The roundover bit can be used to ease sharp edges on furniture, cabinets, crafts, and other woodworking projects. This is a high quality American-made router bit.