Learn More
Grainger Roundover Cut Profile Router Bit 2
Catalog No. 502767878
Encompass_Preferred
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank roundover bit with a 1/4" radius and 1-1/2" cutting length. It is made from solid carbide and has 2 flutes. This bit can be used for decorative edge treatments on wood, laminate and plastic. It has a maximum RPM of 24,000. The overall length of the bit is 3-1/4". This roundover router bit is good for light duty woodworking applications.
Catalog No. 50-276-7878 Supplier Grainger Supplier No. 16Y574
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank roundover bit with a 1/4" radius and 1-1/2" cutting length. It is made from solid carbide and has 2 flutes. This bit can be used for decorative edge treatments on wood, laminate and plastic. It has a maximum RPM of 24,000. The overall length of the bit is 3-1/4". This roundover router bit is good for light duty woodworking applications.