Learn More
Grainger Roundover Cut Profile Router Bit 1
Catalog No. 502766214
Encompass_Preferred
The WESTWARD Roundover Cut Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a roundover profile that cuts rounded edges and corners on wood, plastic, and soft metals. The carbide tipped bit has 2 flutes and a 1/4" shank to fit most common router collets. It can be used for decorative edging, easing sharp edges, and preventing splintering. This USA-made bit is designed for handheld or table-mounted routing at up to 22,000 RPM.
Catalog No. 50-276-6214 Supplier Grainger Supplier No. 16Y536
The WESTWARD Roundover Cut Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a roundover profile that cuts rounded edges and corners on wood, plastic, and soft metals. The carbide tipped bit has 2 flutes and a 1/4" shank to fit most common router collets. It can be used for decorative edging, easing sharp edges, and preventing splintering. This USA-made bit is designed for handheld or table-mounted routing at up to 22,000 RPM.