Learn More
Grainger Roundover Cut Profile Router Bit 1-1/4
Catalog No. 502767009
Encompass_Preferred
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank carbide tipped router bit that creates a rounded edge profile. It has a cutting diameter of 1-1/4", a cutting length of 1/2", and an overall length of 2-1/2". This two flute spiral design router bit is designed for use on wood, plywood, and laminates. It can be used in any standard router and run at speeds up to 24,000 RPM. Made in the USA.
Catalog No. 50-276-7009 Supplier Grainger Supplier No. 16Y535
The WESTWARD Roundover Cut Profile Router Bit is a 1/2" shank carbide tipped router bit that creates a rounded edge profile. It has a cutting diameter of 1-1/4", a cutting length of 1/2", and an overall length of 2-1/2". This two flute spiral design router bit is designed for use on wood, plywood, and laminates. It can be used in any standard router and run at speeds up to 24,000 RPM. Made in the USA.