Learn More
Grainger Router Bit Set 3 Pc Carbide Tipped
Catalog No. 502765225
Encompass_Preferred
The Bosch 15-Piece Router Bit Set includes 15 high speed steel router bits for woodworking applications. The set contains straight, roundover, cove, chamfer, dovetail, and roman ogee router bits in 1/4", 3/8", and 1/2" shank diameters. These carbide-tipped router bits feature precision ground cutters for clean cuts in wood, plywood, and laminate. The bits have a bright finish to resist corrosion. This router bit set comes in a plastic case for convenient storage and organization.
Catalog No. 50-276-5225 Supplier Grainger Supplier No. 16Y598
The Bosch 15-Piece Router Bit Set includes 15 high speed steel router bits for woodworking applications. The set contains straight, roundover, cove, chamfer, dovetail, and roman ogee router bits in 1/4", 3/8", and 1/2" shank diameters. These carbide-tipped router bits feature precision ground cutters for clean cuts in wood, plywood, and laminate. The bits have a bright finish to resist corrosion. This router bit set comes in a plastic case for convenient storage and organization.