Learn More
Grainger Router Bit Set 30 Pc Carbide Tipped
Catalog No. 502765082
Encompass_Preferred
The Bosch Fractional Router Bit Set contains 12 router bits made of high-speed steel with 1/4-inch shanks. The set includes slotting, straight, V-groove, roundover, cove, dovetail, and chamfer router bits with diameters ranging from 1/4 to 1 inch. The bits are designed for use in handheld or table-mounted routers and are suited for tasks like edge forming, hinge mortising, slot cutting, and decorative profiling on wood, laminate, and plastic. This router bit set comes in a plastic case for convenient storage and transport.
Catalog No. 50-276-5082 Supplier Grainger Supplier No. 16Y605
The Bosch Fractional Router Bit Set contains 12 router bits made of high-speed steel with 1/4-inch shanks. The set includes slotting, straight, V-groove, roundover, cove, dovetail, and chamfer router bits with diameters ranging from 1/4 to 1 inch. The bits are designed for use in handheld or table-mounted routers and are suited for tasks like edge forming, hinge mortising, slot cutting, and decorative profiling on wood, laminate, and plastic. This router bit set comes in a plastic case for convenient storage and transport.