Learn More
Grainger Router Bit Set 4 Pc Carbide Tipped
Catalog No. 502765586
Encompass_Preferred
The DeWalt DW6913 router bit set contains 15 high speed steel router bits in 1/4", 3/8", and 1/2" shank sizes. The set includes straight, roundover, cove, V-groove, chamfer, dovetail, and Roman Ogee bits as well as a T-slot cutter, flush trim bit, and two edge guide bits. The 1/4" shank bits can be used in laminate trimmers and compact routers while the larger shank sizes fit full size routers. All bits feature precision ground carbide tips for clean cuts in wood, plastic, and soft metals. The bits come packed in a molded case.
Catalog No. 50-276-5586 Supplier Grainger Supplier No. 16Y603
The DeWalt DW6913 router bit set contains 15 high speed steel router bits in 1/4", 3/8", and 1/2" shank sizes. The set includes straight, roundover, cove, V-groove, chamfer, dovetail, and Roman Ogee bits as well as a T-slot cutter, flush trim bit, and two edge guide bits. The 1/4" shank bits can be used in laminate trimmers and compact routers while the larger shank sizes fit full size routers. All bits feature precision ground carbide tips for clean cuts in wood, plastic, and soft metals. The bits come packed in a molded case.