Learn More
Grainger Router Corded 3.25 hp
Catalog No. 502749188
Encompass_Preferred
The FisherbrandTM 502749188 is a polypropylene microcentrifuge tube rack that holds 96 1.5mL tubes in an 8x12 array. It is made of autoclavable polypropylene and has alphanumeric grid references for easy sample identification. The rack measures 127x86x45mm (5x3.4x1.8in) and features a detachable lid with condensation rings to prevent sample cross-contamination. This microcentrifuge tube rack provides an organized storage solution for 1.5mL tubes during centrifugation and prevents tube damage. It can withstand temperatures from -20°C to 121°C and is compatible with most microcentrifuge rotors. The sturdy one-piece construction ensures rack stability when loaded.
Catalog No. 50-274-9188 Supplier Grainger Supplier No. 14F183
The FisherbrandTM 502749188 is a polypropylene microcentrifuge tube rack that holds 96 1.5mL tubes in an 8x12 array. It is made of autoclavable polypropylene and has alphanumeric grid references for easy sample identification. The rack measures 127x86x45mm (5x3.4x1.8in) and features a detachable lid with condensation rings to prevent sample cross-contamination. This microcentrifuge tube rack provides an organized storage solution for 1.5mL tubes during centrifugation and prevents tube damage. It can withstand temperatures from -20°C to 121°C and is compatible with most microcentrifuge rotors. The sturdy one-piece construction ensures rack stability when loaded.