Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Routine DNA Ladder

Catalog No. p-3016192 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Bands:
11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Bands
BP2581200 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2581200

Ready-to-use DNA size standard is room-temperature stable for up to two years

  • Includes a landmark doublet at 1400 and 1550 bp

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Form Liquid
For Use With (Application) PCR size confirmation
No. of Bands 11
Quantity 1000 μL
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.