Learn More
StatLab Routine Embedding Cassettes
SDP
Catalog No. 21403480
Change view
Click to view available options
Color:
White
Catalog No. Color
21-403-480 White
1 options
Catalog No. 21-403-480 Supplier StatLab Supplier No. M4852
Add to Cart
Add to Cart
Histosette II White cassettes.
  • Unique front-hinged design with a rear tab that allows for unobstructed access to the front labeling surface
  • Lids separate, bulk packed
  • 1000/Cs.
Specifications
Color White
Product Type Embedding Cassettes