Learn More
Medchemexpress LLC HY-109073 5mg Medchemexpress, Rovazolac CAS:1454288-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090735MG
Encompass_Preferred
Medchemexpress, HY-109073 5mg Rovazolac CAS:1454288-88-0 Rovazolac is a liver x receptor (LXR) modulator extracted from patent WO2013130892A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8666