Learn More
Abcam 5(6)-Carbo x y- x -rhodamine, 25MG
Catalog No. 5051302
Encompass_Preferred
MW 1069.2 Da, Purity >90%. Stable, long wavelength, water soluble fluorophore has found widespread use in DNA-labeling, cell staining and protein labeling applications.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-513-02 Supplier Abcam Supplier No. AB27536025MG
MW 1069.2 Da, Purity >90%. Stable, long wavelength, water soluble fluorophore has found widespread use in DNA-labeling, cell staining and protein labeling applications.
The product is subject to the following: Abcam Restricted Use Statement