Learn More
Aat Bioquest ROX DYE QPCR CALIB 100UL
SDP
Catalog No. NC3640512
Encompass_Preferred
  • Liquid form for easy handling
  • 10,000X concentration for efficient calibration
  • 100 µL unit size suitable for multiple uses
  • Designed for real-time thermal cycler calibration
  • Compatible with ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems
  • Recommended for use with PCR microplate
  • ROX filter specification for optimal measurement
  • Excitation wavelength of 578 nm for accurate detection
  • Emission wavelength of 595 nm for enhanced signal
  • Water as the solvent ensures stability
  • Store by freezing (< -15 °C) and minimizing light exposure
  • Calibration frequency every six months or as needed
  • Intended for research use only (RUO)
Catalog No. NC3640512 Supplier Aat Bioquest Supplier No. 67034
  • Liquid form for easy handling
  • 10,000X concentration for efficient calibration
  • 100 µL unit size suitable for multiple uses
  • Designed for real-time thermal cycler calibration
  • Compatible with ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems
  • Recommended for use with PCR microplate
  • ROX filter specification for optimal measurement
  • Excitation wavelength of 578 nm for accurate detection
  • Emission wavelength of 595 nm for enhanced signal
  • Water as the solvent ensures stability
  • Store by freezing (< -15 °C) and minimizing light exposure
  • Calibration frequency every six months or as needed
  • Intended for research use only (RUO)