Learn More
Biotium ROX Passive Reference Dye
Catalog No. 501964454
Encompass_Preferred
ROX reference dye is used to normalize well-to-well variations that may occur during quantitative, real-time PCR reactions. The dye is supplied as a 25 uM solution in 10 mM Tris-HCL (pH 8.6), 0.1 mM EDTA, and 0.01% Tween-20.
Catalog No. 50-196-4454 Supplier Biotium Supplier No. 29052
ROX reference dye is used to normalize well-to-well variations that may occur during quantitative, real-time PCR reactions. The dye is supplied as a 25 uM solution in 10 mM Tris-HCL (pH 8.6), 0.1 mM EDTA, and 0.01% Tween-20.