Learn More
As One International Inc ROX internal reference dye, 3 x 0.2 ml
Catalog No. 501656012
Encompass_Preferred
A passive reference dye to compensate for non-PCR related variations in the fluorescence. The fluorescence from the passive reference dye does not change during the course of the PCR reaction but provides a stable baseline to which samples are normalized. The excitation and emission of the reference dye are 584 nm and 612 nm, respectively.
Catalog No. 50-165-6012 Supplier As One International Inc Supplier No. AO351513
A passive reference dye to compensate for non-PCR related variations in the fluorescence. The fluorescence from the passive reference dye does not change during the course of the PCR reaction but provides a stable baseline to which samples are normalized. The excitation and emission of the reference dye are 584 nm and 612 nm, respectively.