Learn More
Medchemexpress LLC HY-13426 100mg Medchemexpress, Roxadustat CAS:808118-40-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13426100MG
Encompass_Preferred
Medchemexpress, HY-13426 100mg Roxadustat CAS:808118-40-3 Roxadustat is an oral hypoxia-inducible factor prolyl-hydroxylase inhibitor (HIF-PHI) that promotes erythropoiesis through increasing endogenous erythropoietin, improving iron regulation, and reducing hepcidin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8992