Learn More
Selleck Chemical LLC Roxadustat (FG-4592) 10mM/1mL 808118-40-3 ASP1517
SDP
Catalog No. 501360851
Encompass_Preferred
Roxadustat (FG-4592) is an HIF-? prolyl hydroxylase inhibitor in a cell-free assay, stabilizes HIF-2 and induces EPO production. Roxadustat potentiates RSL3 induced ferroptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0851 Supplier Selleck Chemical LLC Supplier No. S100710MM/1ML
Roxadustat (FG-4592) is an HIF-? prolyl hydroxylase inhibitor in a cell-free assay, stabilizes HIF-2 and induces EPO production. Roxadustat potentiates RSL3 induced ferroptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.