Learn More
Medchemexpress LLC Roxatidine | 78273-80-0 | 98.25% | 306.40 | 50 MG
SDP
Catalog No. 5000350315
Encompass_Preferred
Roxatidine is an active metabolite of Roxatidine acetate hydrochloride and an orally active histamine H2-receptor antagonist. It is an anti-ulcer agent that suppresses histamine release, inhibits proton secretion, and inhibits the production of VEGF-1. This agent also demonstrates anti-allergic inflammatory effects and is promising for research into gastric and duodenal ulcers.
  • Orally active histamine H2-receptor antagonist
  • Suppresses histamine release and inhibits proton secretion
  • Inhibits production of VEGF-1
  • Exhibits anti-allergic inflammatory effects
  • Potential for research into gastric and duodenal ulcers
  • Suppresses pro-inflammatory cytokine production and signaling pathways (in vitro)
  • Inhibits inflammatory mediators such as PGE2, NO, and histamine (in vitro)
  • Ameliorates allergic hypersensitivity and inflammation in anaphylactic animal models (in vivo)
Catalog No. 50-003-50315 Supplier Medchemexpress LLC Supplier No. HY13794150MG
Add to Cart
Add to Cart
Roxatidine is an active metabolite of Roxatidine acetate hydrochloride and an orally active histamine H2-receptor antagonist. It is an anti-ulcer agent that suppresses histamine release, inhibits proton secretion, and inhibits the production of VEGF-1. This agent also demonstrates anti-allergic inflammatory effects and is promising for research into gastric and duodenal ulcers.
  • Orally active histamine H2-receptor antagonist
  • Suppresses histamine release and inhibits proton secretion
  • Inhibits production of VEGF-1
  • Exhibits anti-allergic inflammatory effects
  • Potential for research into gastric and duodenal ulcers
  • Suppresses pro-inflammatory cytokine production and signaling pathways (in vitro)
  • Inhibits inflammatory mediators such as PGE2, NO, and histamine (in vitro)
  • Ameliorates allergic hypersensitivity and inflammation in anaphylactic animal models (in vivo)