Learn More
Medchemexpress LLC Roxatidine | 78273-80-0 | 98.25% | 306.41 | 100 MG
SDP
Catalog No. 5000350542
Encompass_Preferred
Roxatidine is an active metabolite of Roxatidine acetate hydrochloride and an orally active histamine H2-receptor antagonist. As an anti-ulcer agent, it suppresses histamine release, inhibiting proton secretion, and inhibits the production of VEGF-1, which is a marker of inflammation and angiogenesis. It also has an anti-allergic inflammatory effect and is being researched for gastric and duodenal ulcers.
  • Orally active histamine H2-receptor antagonist
  • Suppresses histamine release (inhibiting proton secretion)
  • Inhibits the production of VEGF-1
  • Exhibits anti-allergic inflammatory effects
  • Researched for gastric and duodenal ulcers
Catalog No. 50-003-50542 Supplier Medchemexpress LLC Supplier No. HY137941100MG
Add to Cart
Add to Cart
Roxatidine is an active metabolite of Roxatidine acetate hydrochloride and an orally active histamine H2-receptor antagonist. As an anti-ulcer agent, it suppresses histamine release, inhibiting proton secretion, and inhibits the production of VEGF-1, which is a marker of inflammation and angiogenesis. It also has an anti-allergic inflammatory effect and is being researched for gastric and duodenal ulcers.
  • Orally active histamine H2-receptor antagonist
  • Suppresses histamine release (inhibiting proton secretion)
  • Inhibits the production of VEGF-1
  • Exhibits anti-allergic inflammatory effects
  • Researched for gastric and duodenal ulcers