Learn More
Medchemexpress LLC Roxatidine | 78273-80-0 | MFCD00871807 | 98.3% | 306.41 g/mol | C17H26N2O3 | 5 MG
SDP
Catalog No. 5000222776
Encompass_Preferred
Roxatidine is a histamine H2 receptor antagonist supplied as a research chemical for in vitro and pharmacological studies. It suppresses histamine-stimulated gastric acid secretion and has reported anti-inflammatory activity. The compound is provided as a crystalline powder and as a DMSO solution for laboratory use; it is intended for research use only.
  • Histamine H2 receptor antagonist
  • Used in studies of gastric acid secretion and ulcer biology
  • Reported anti-inflammatory activity mediated via NF-κB and MAPK pathways
  • Available as powder and as a 10 mM DMSO solution
  • High purity (approximately 98.3%)
  • Molecular weight 306.41 g/mol; formula C17H26N2O3
  • For research use only; not for human or veterinary use
Catalog No. 50-002-22776 Supplier Medchemexpress LLC Supplier No. HY1379415MG
Roxatidine is a histamine H2 receptor antagonist supplied as a research chemical for in vitro and pharmacological studies. It suppresses histamine-stimulated gastric acid secretion and has reported anti-inflammatory activity. The compound is provided as a crystalline powder and as a DMSO solution for laboratory use; it is intended for research use only.
  • Histamine H2 receptor antagonist
  • Used in studies of gastric acid secretion and ulcer biology
  • Reported anti-inflammatory activity mediated via NF-κB and MAPK pathways
  • Available as powder and as a 10 mM DMSO solution
  • High purity (approximately 98.3%)
  • Molecular weight 306.41 g/mol; formula C17H26N2O3
  • For research use only; not for human or veterinary use