Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Roxatidine | 78273-80-0 | MFCD00871807 | 98.3% | C17H26N2O3 | 10MG
SDP

Catalog No. 5000261303
Encompass_Preferred

Roxatidine is a histamine H2-receptor antagonist and the active metabolite of roxatidine acetate hydrochloride, supplied as a research standard for studies of gastric acid secretion, peptic ulcers, and inflammatory responses. The material is provided as a solid powder for in vitro and preclinical research.

  • High purity (~98.3%) suitable for research applications
  • Confirmed identity: CAS 78273-80-0
  • Molecular formula C17H26N2O3; mol wt 306.41 g/mol
  • Soluble in DMSO for assay preparation
  • Available in small milligram quantities for screening and method development

Catalog No. 50-002-61303 Supplier Medchemexpress LLC Supplier No. HY13794110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Roxatidine is a histamine H2-receptor antagonist and the active metabolite of roxatidine acetate hydrochloride, supplied as a research standard for studies of gastric acid secretion, peptic ulcers, and inflammatory responses. The material is provided as a solid powder for in vitro and preclinical research.

  • High purity (~98.3%) suitable for research applications
  • Confirmed identity: CAS 78273-80-0
  • Molecular formula C17H26N2O3; mol wt 306.41 g/mol
  • Soluble in DMSO for assay preparation
  • Available in small milligram quantities for screening and method development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.