Learn More
Medchemexpress LLC Roxatidine acetate | 78628-28-1 | 5 G
SDP
Catalog No. 5000405468
Encompass_Preferred
Roxatidine acetate is a potent, selective, competitive, and orally active histamine H2-receptor antagonist. It demonstrates antisecretory potency against gastric acid secretion and can suppress inflammatory responses. This compound is suitable for research into gastric and duodenal ulcers, and has shown antitumor activity.
  • Potent, selective, competitive, and orally active histamine H2-receptor antagonist
  • Suppresses gastric acid secretion
  • Inhibits inflammatory responses
  • Useful for gastric and duodenal ulcer research
  • Exhibits antitumor activity
  • Attenuates mast cell-mediated allergic inflammation
Catalog No. 50-004-05468 Supplier Medchemexpress LLC Supplier No. HYB0305A5G
Roxatidine acetate is a potent, selective, competitive, and orally active histamine H2-receptor antagonist. It demonstrates antisecretory potency against gastric acid secretion and can suppress inflammatory responses. This compound is suitable for research into gastric and duodenal ulcers, and has shown antitumor activity.
  • Potent, selective, competitive, and orally active histamine H2-receptor antagonist
  • Suppresses gastric acid secretion
  • Inhibits inflammatory responses
  • Useful for gastric and duodenal ulcer research
  • Exhibits antitumor activity
  • Attenuates mast cell-mediated allergic inflammation