Learn More
Apexbio Technology LLC Roxatidine Acetate HCl 93793-83-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063382
Encompass_Preferred
Roxatidine Acetate HCl (CAS 93793-83-0) is a selective competitive antagonist of histamine H2 receptors By occupying these receptors it attenuates histamine-induced gastric acid secretion in parietal cells Roxatidine Acetate HCl is primarily utilized in biomedical studies investigating gastric physiology and acid-related gastrointestinal disorders Its receptor-specific inhibitory properties make it valuable in research exploring histaminergic signaling pathways and related pharmacological interventions
Catalog No. 50-000-63382 Supplier Apexbio Technology LLC Supplier No. B15655.1
Roxatidine Acetate HCl (CAS 93793-83-0) is a selective competitive antagonist of histamine H2 receptors By occupying these receptors it attenuates histamine-induced gastric acid secretion in parietal cells Roxatidine Acetate HCl is primarily utilized in biomedical studies investigating gastric physiology and acid-related gastrointestinal disorders Its receptor-specific inhibitory properties make it valuable in research exploring histaminergic signaling pathways and related pharmacological interventions