Learn More
Selleck Chemical LLC Roxatidine Acetate HCl S1880-50mg
SDP
Catalog No. 503110799
Encompass_Preferred
Roxatidine Acetate HCl(HOE 760) is a specific and competitive histamin H2-receptor antagonist with IC50 of 3 2 M inhibits gastric acid secretion and ulcer formation
Catalog No. 50-311-0799 Supplier Selleck Chemical LLC Supplier No. S188050MG
Roxatidine Acetate HCl(HOE 760) is a specific and competitive histamin H2-receptor antagonist with IC50 of 3 2 M inhibits gastric acid secretion and ulcer formation